ContactUsLinkComponent
Powerful, compact and energy-efficient recirculating chiller for cooling applications up to -30 C. The on-demand and speed-controlled compressor, operated with natural refrigerant, achieves a cooling capacity of 800 W at minimum energy consumption 1 * 1 pezzi
Codice catalogo IKAA25006638
Nome del fornitore:  IKA
Powerful, compact and energy-efficient recirculating chiller for cooling applications up to -30 C. The on-demand and speed-controlled compressor, operated with natural refrigerant, achieves a cooling capacity of 800 W at minimum energy consumption 1 * 1 pezzi
Codice catalogo IKAA25006638
Nome del fornitore:  IKA
Frequently Bought Together