Plastic replacement tips
Plastic replacement tips for forceps, curved, very fine, Acciaio inossidabile, 40 mm, Ø 0,5 mm
Codice catalogo FSCI11988-03
Marketsource
Nome del fornitore: FINE SCIENCE TOOLS
Plastic replacement tips
Codice catalogo FSCI11988-03
Nome del fornitore: FINE SCIENCE TOOLS
Marketsource
Vai a
Product Details & Documents
Pack of (2) PVDF tips and (3) screws. Tips are made of polyvinylidene fluoride (PVDF) and are electrostatic discharge (ESD) safe.
Compatible with Item No. 11980-13.
Specifiche
- Descrizione:Plastic replacement tips for forceps, curved, very fine
- Lunghezza:40 mm
- Materiale:Acciaio inossidabile
- Tip size:Ø 0,5 mm