Hook2Loop Bungee tool tether, DBI-SALA®, 3M™
Hook2Loop Bungee tether, yellow, 4,5 kg capacity
Codice catalogo CPSD1500048
Nome del fornitore: CAPITAL SAFETY
Hook2Loop Bungee tool tether, DBI-SALA®, 3M™
Codice catalogo CPSD1500048
Nome del fornitore: CAPITAL SAFETY
Vai a
Product Details & Documents
Bungee tool tether with self-locking carabiner.
- Adjusts to virtually any wrist size with hook and loop system
- Replaceable steel cable retractor fits into a sewn-in pocket and allows tools to easily be tied off using an attached thumb-operated trigger
- Elastic band helps ensure a comfortable fit
- Attached trigger can be used to quickly work with many hand tools
This is a very versatile extension that can be used with virtually any tool, such as spud wrenches, scaffold wrenches, adjustable wrenches, and more.
Designed for use in general industrial, oil and gas, mining, construction, and transportation industries.
Documents
3M™ Fall Protection Core Product Range 3M™ Fall Protection for Tools
Product Literature
Specifiche
- Descrizione:Hook2Loop Bungee tether, yellow, 4,5 kg capacity
- Seq:01