Orders LinkContactUsLinkComponent
Accessories for PLC purification systems
Accessories for PLC purification systems
Rack, code 29, holds quantity of 60, 12/13×75 mm tubes ( 5/6 ml)
Catalog # GILI150429
Supplier:   GILSON LCMS
Accessories for PLC purification systems
Catalog # GILI150429
Supplier:  GILSON LCMS
Accessories for PLC purification systems
Rack, code 29, holds quantity of 60, 12/13×75 mm tubes ( 5/6 ml)
Price
SFr. 100.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Descrizione:
    Rack, code 29, holds quantity of 60, 12/13×75 mm tubes ( 5/6 ml)
  • Seq:
    43