Orders LinkContactUsLinkComponent
PLC purification systems
PLC purification systems
PLC 2250 purification system, 5 - 250 ml/min. flow rate, 230 bar max. pressure
Catalog # GILI21140002
Supplier:   GILSON LCMS
PLC purification systems
Catalog # GILI21140002
Supplier:  GILSON LCMS
PLC purification systems
PLC 2250 purification system, 5 - 250 ml/min. flow rate, 230 bar max. pressure
Price
SFr. 43’420.00
Log in for availability
1 * 1 ST
Product Details & Documents
PLC 2050/2250/2500 purification systems are designed for preparative liquid chromatography (PLC) to aspirate mobile phase solvents, form gradients, inject liquid samples, and provide solutions for a preparative column for high performance liquid chromatography (HPLC), centrifugal partition chromatography (CPC), or counter-current chromatography (CCC) applications.
  • PLC 2050: Flow rates up to 50 ml/min with pressure up to 300 bar
  • PLC 2250: Flow rates up to 250 ml/min with pressure up to 230 bar
  • PLC 2500: Flow rates up to 500 ml/min with pressure up to 110 bar
  • Intuitive touchscreen interface for easy method writing and modyfying of parameters
  • Autosampler available for automated sample injections for increased throughput
  • Costs: 'All in one' system
  • Safety: Compact footprint that fits into a fume hood
With the capability to purify compounds by preparative HPLC and flash chromatography on the same instrument, Gilson's PLC 2050/2250/2500 purification systems simplify and streamline compound purification. These compact systems feature UV/VIS detection with an optional upgrade to a diode array detector (DAD) or evaporative light scattering detection (ELSD), an integrated pumping system, fraction collector, touchscreen control, and Gilson glider software. These customisable instruments are available in simple configurations for basic applications, but with a wide range of upgrade options to meet your lab’s demands.
Electrical: 110 to 120 / 220 to 240 V
Specifications
  • Descrizione:
    PLC 2250 purification system, 5 - 250 ml/min. flow rate, 230 bar max. pressure
  • Seq:
    02
Accessories
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040023
    Rack set of 3 racks, 25×150 mm tubes
    Log in for availability
    1 * 3 ST - SFr. 1’370.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040024
    Rack set of 3 racks, 28×150 mm tubes
    Log in for availability
    1 * 3 ST - SFr. 1’370.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040025
    Rack set of 3 racks, 29,5×200 mm tubes
    Log in for availability
    1 * 3 ST - SFr. 1’370.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040026
    Rack set of 3 racks, 13×73 mm hemo tubes
    Log in for availability
    1 * 3 ST - SFr. 1’370.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040027
    Rack funnel, 16 outlets ¹/₄" Ext.Ø
    Log in for availability
    1 * 3 ST - SFr. 2’550.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040036
    Sample loop, 40 ml auto, ¹/₈" Ext.Ø
    Log in for availability
    1 * 1 ST - SFr. 761.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040037
    Sample loop, 50 ml auto, ¹/₈" Ext.Ø
    Log in for availability
    1 * 1 ST - SFr. 915.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040181
    Manual injection valve with tubing kit (PLC 2050)
    Log in for availability
    1 * 1 ST - SFr. 2’830.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040182
    Manual injection valve with tubing kit (PLC 2250/2500)
    Log in for availability
    1 * 1 ST - SFr. 2’830.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040184
    Injection pump Ext (50 ml/min, 300 Bar)
    Log in for availability
    1 * 1 ST - SFr. 14’910.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040191
    Rack set of 3 racks, 12×65 mm tubes
    Log in for availability
    1 * 3 ST - SFr. 1’370.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040019
    Rack set of 3 racks, 18×150/180 mm tubes
    Log in for availability
    1 * 3 ST - SFr. 978.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040020
    Rack set of 3 racks, 13×100 mm tubes
    Log in for availability
    1 * 3 ST - SFr. 1’370.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040021
    Rack set of 3 racks, 16×150 mm tubes
    Log in for availability
    1 * 3 ST - SFr. 1’370.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040022
    Rack set of 3 racks, 21×150 mm tubes
    Log in for availability
    1 * 3 ST - SFr. 1’370.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040028
    3 Rack overlays for 18 mm tubes
    Log in for availability
    1 * 3 ST - SFr. 153.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040029
    3 Rack overlays 13 mm tubes
    Log in for availability
    1 * 3 ST - SFr. 153.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040030
    Sample loop, 1 ml auto, ¹/₈" Ext.Ø
    Log in for availability
    1 * 1 ST - SFr. 133.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040031
    Sample loop, 2 ml auto, ¹/₈" Ext.Ø
    Log in for availability
    1 * 1 ST - SFr. 143.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040032
    Sample loop, 5 ml auto, ¹/₈" Ext.Ø
    Log in for availability
    1 * 1 ST - SFr. 239.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040033
    Sample loop, 10 ml auto, ¹/₈" Ext.Ø
    Log in for availability
    1 * 1 ST - SFr. 315.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040034
    Sample loop, 20 ml auto, ¹/₈" Ext.Ø
    Log in for availability
    1 * 1 ST - SFr. 519.00
Product Information
  • Catalog #
    Descrizione
    Availability
    Price
  • GILI21040035
    Sample loop, 30 ml auto, ¹/₈" Ext.Ø
    Log in for availability
    1 * 1 ST - SFr. 607.00