Orders LinkContactUsLinkComponent
Fraction collectors
Fraction collectors
Fraction collector FC204 with low pressure valve, HPLC purification technique, 110 - 120/220 - 240 V
Codice catalogo GILI171041
Nome del fornitore:   GILSON LCMS
Fraction collectors
Codice catalogo GILI171041
Nome del fornitore:  GILSON LCMS
Fraction collectors
Fraction collectors
Fraction collector FC204 with low pressure valve, HPLC purification technique, 110 - 120/220 - 240 V
Price
8 078.40 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Ideal solutions for small to large capacity collection. Gilson offers advanced solutions for fraction collection with a complete line of stand‑alone and software-controlled fraction collectors ranging from small footprint to large vessel capacity models. The built-in, easy-to-use keypad makes setup and operation simple and capable of interfacing with any HPLC system or control the system through your PC with TRILUTION® LC software.
  • Collect fractions by time, drop, or peak to suit all your LC applications
  • Choose the compact FC 203B for small-capacity collection or the FC 204 for large volumes and capacities
  • In peak mode, collectors monitor a detector signal and identify peaks using either an adaptive-slope algorithm or a specified millivolt threshold
  • Time: Up to 10 programmable time windows can be added for collecting only what you want, while discarding the column’s void volume, peaks of no interest, and equilibration volumes
  • 3-way diverter valve prevents contamination of collected fractions (optional)
  • Compact size fits in most fume hoods
FC203B and FC203B with valve: choose among peak, drop, or time collection options with the FC 203B fraction collector. In peak mode, the FC 203B monitors a detector signal and identifies peaks using either an adaptive-slope algorithm or a specified millivolt threshold. With up to ten programmable time windows to use, collect only what you want and discard the column void volume, peaks of no interest, and equilibration volumes. Multicolumn adapters are available for collecting up to 18 separate channels and an optional 3-way diverter valve prevents contamination of collected fractions.
FC204: Collect samples by peak, drop, or time using the FC 204 Fraction Collector. Add up to ten programmable time windows to collect only what you want, eliminating void volumes, equilibration volumes, and peaks of no interest. In peak mode, the collector monitors a detector signal, identifying peaks using an adaptive-slope algorithm or a specified millivolt threshold. The 3-way diverter valve prevents contamination of fractions. Multicolumn adapters allow for collection from up to 18 separate channels.
Specifiche
  • Descrizione:
    Fraction collector FC204 with low pressure valve, HPLC purification technique, 110 - 120/220 - 240 V
  • Seq:
    03
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 24, holds quantity of 14, 28×57 mm scintilation vials (20 ml)
    Rack, code 24, holds quantity of 14, 28×57 mm scintilation vials (20 ml)
    GILI150427
    Log in per disponibilità
    1 * 1 ST - SFr. 92.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 28, holds quantity of 108 of 10×75 mm tubes (3,5 ml)
    Rack, code 28, holds quantity of 108 of 10×75 mm tubes (3,5 ml)
    GILI150420
    Log in per disponibilità
    1 * 1 ST - SFr. 91.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 21, holds quantity of 60, 13×100 mm tubes (9 ml)
    Rack, code 21, holds quantity of 60, 13×100 mm tubes (9 ml)
    GILI150422
    Log in per disponibilità
    1 * 1 ST - SFr. 103.40
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 22, holds quantity of 44, 18×150 mm tubes (25 ml), for use with FC204
    Rack, code 22, holds quantity of 44, 18×150 mm tubes (25 ml), for use with FC204
    GILI150424
    Log in per disponibilità
    1 * 1 ST - SFr. 200.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 20, holds quantity of 108, 10×100 mm tubes (4,5 ml)
    Rack, code 20, holds quantity of 108, 10×100 mm tubes (4,5 ml)
    GILI150425
    Log in per disponibilità
    1 * 1 ST - SFr. 142.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 23, holds quantity of 44, 17×55 mm scintilation vials (6,8 ml)
    Rack, code 23, holds quantity of 44, 17×55 mm scintilation vials (6,8 ml)
    GILI150426
    Log in per disponibilità
    1 * 1 ST - SFr. 92.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 29, holds quantity of 60, 12/13×75 mm tubes ( 5/6 ml)
    Rack, code 29, holds quantity of 60, 12/13×75 mm tubes ( 5/6 ml)
    GILI150429
    Log in per disponibilità
    1 * 1 ST - SFr. 100.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 22U, Universal, holds 10 - 18×100 - 175 mm tubes (4 ml), for use with FC204
    Rack, code 22U, Universal, holds 10 - 18×100 - 175 mm tubes (4 ml), for use with FC204
    GILI150498
    Log in per disponibilità
    1 * 1 ST - SFr. 357.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 28, holds quantity of 108, 10×75 mm tubes (5/pack)
    Rack, code 28, holds quantity of 108, 10×75 mm tubes (5/pack)
    GILI152420
    Log in per disponibilità
    1 * 5 ST - SFr. 405.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 21, holds quantity of 60, 13×100 mm tubes (9 ml) (5/pack)
    Rack, code 21, holds quantity of 60, 13×100 mm tubes (9 ml) (5/pack)
    GILI152422
    Log in per disponibilità
    1 * 5 ST - SFr. 357.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 22, holds quantity of 44, 18×150 mm tubes (25 ml) (5/pack), for use with FC204
    Rack, code 22, holds quantity of 44, 18×150 mm tubes (25 ml) (5/pack), for use with FC204
    GILI152424
    Log in per disponibilità
    1 * 5 ST - SFr. 964.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 23, holds quantity of 44, 17×55 scintilation vials (5/pack)
    Rack, code 23, holds quantity of 44, 17×55 scintilation vials (5/pack)
    GILI152426
    Log in per disponibilità
    1 * 5 ST - SFr. 430.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 24, holds quantity of 14, 28×57 mm scintilation vials (5/pack)
    Rack, code 24, holds quantity of 14, 28×57 mm scintilation vials (5/pack)
    GILI152427
    Log in per disponibilità
    1 * 5 ST - SFr. 426.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 23W, holds quantity of 44, 15×45 mm WISP vials (4 ml)
    Rack, code 23W, holds quantity of 44, 15×45 mm WISP vials (4 ml)
    GILI270433
    Log in per disponibilità
    1 * 1 ST - SFr. 282.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 11, holds quantity of 105, 13×100 mm, 9 ml, for use with fraction collectors
    Rack, code 11, holds quantity of 105, 13×100 mm, 9 ml, for use with fraction collectors
    GILI170413
    Log in per disponibilità
    1 * 1 ST - SFr. 1’230.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 29LE, holds quantity of 60, 1,5 ml microcentrifuge tubes
    Rack, code 29LE, holds quantity of 60, 1,5 ml microcentrifuge tubes
    GILI2704342
    Log in per disponibilità
    1 * 1 ST - SFr. 351.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 29SE, holds quantity of 60, 0,5 ml microcentrifuge tubes
    Rack, code 29SE, holds quantity of 60, 0,5 ml microcentrifuge tubes
    GILI2704341
    Log in per disponibilità
    1 * 1 ST - SFr. 428.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 4, holds quantity of 80, 12×75 mm tubes, 5 ml, for use with fraction collectors
    Rack, code 4, holds quantity of 80, 12×75 mm tubes, 5 ml, for use with fraction collectors
    GILI130412
    Log in per disponibilità
    1 * 1 ST - SFr. 681.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 14, holds quantity of 128, 12×75 or 100 mm, 5 ml, for use with fraction collectors
    Rack, code 14, holds quantity of 128, 12×75 or 100 mm, 5 ml, for use with fraction collectors
    GILI170414
    Log in per disponibilità
    1 * 1 ST - SFr. 970.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 0, holds quantity of 80, 12×32 mm vials, 2 ml, for use with fraction collectors
    Rack, code 0, holds quantity of 80, 12×32 mm vials, 2 ml, for use with fraction collectors
    GILI270430
    Log in per disponibilità
    1 * 1 ST - SFr. 65.40
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • 3-way valve kit with 1,5 mm Int.Ø, FC203, for use with fraction collectors
    3-way valve kit with 1,5 mm Int.Ø, FC203, for use with fraction collectors
    GILI170730
    Log in per disponibilità
    1 * 1 ST - SFr. 898.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • FC-204 Prep valve assembly package, for use with fraction collectors
    FC-204 Prep valve assembly package, for use with fraction collectors
    GILI170735
    Log in per disponibilità
    1 * 1 ST - SFr. 1’630.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 15, holds 1×96-well microplate, for use with fraction collectors
    Rack, code 15, holds 1×96-well microplate, for use with fraction collectors
    GILI170415
    Log in per disponibilità
    1 * 1 ST - SFr. 114.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 16, holds 4×96-well microplates, for use with fraction collectors
    Rack, code 16, holds 4×96-well microplates, for use with fraction collectors
    GILI170416
    Log in per disponibilità
    1 * 1 ST - SFr. 241.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack heightener, code 17, for use with fraction collectors
    Rack heightener, code 17, for use with fraction collectors
    GILI170417
    Log in per disponibilità
    1 * 1 ST - SFr. 221.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 17, microplate, for use with fraction collectors
    Rack, code 17, microplate, for use with fraction collectors
    GILI170418
    Log in per disponibilità
    1 * 1 ST - SFr. 424.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • 3-way valve kit with 1,5 mm Int.Ø, FC204, for use with fraction collectors
    3-way valve kit with 1,5 mm Int.Ø, FC204, for use with fraction collectors
    GILI170734
    Log in per disponibilità
    1 * 1 ST - SFr. 999.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Rack, code 16D, holds 4 deep well microplates, for use with fraction collectors
    Rack, code 16D, holds 4 deep well microplates, for use with fraction collectors
    GILI170416D
    Log in per disponibilità
    1 * 1 ST - SFr. 241.00
Informazioni sul prodotto
  • Codice catalogo
    Disponibilità
    Price
  • Multiple column adapter, for use with fraction collectors
    Multiple column adapter, for use with fraction collectors
    GILI174732
    Log in per disponibilità
    1 * 1 ST - SFr. 885.00