Iron AAS Standards, (CRMs), Fluka™
Ferro, Fe, 10000 mg/L, 5% HCl, Plastic bottle, 100 mL
Codice catalogo HONCE4110-100ML
Nome del fornitore: Honeywell Chemicals
Iron AAS Standards, (CRMs), Fluka™
Codice catalogo HONCE4110-100ML
Nome del fornitore: Honeywell Chemicals
Numero CAS:
Vai a
Product Details & Documents
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Specifiche
Specifiche
- Concentrazione:10000 mg/L
- Matrice:5% HCl
- Tipo di conf.:Plastic bottle
- Conf:100 mL
- Componenti:Ferro
- Simbolo:Fe
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



