Iron AAS Standards, (CRMs), Fluka™
Ferro, Fe, 1000 mg/L, 2% HCl, Plastic bottle, 500 mL
HONCE4027-500ML
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
- Concentrazione:1000 mg/L
- Matrice:2% HCl
- Tipo di conf.:Plastic bottle
- Conf:500 mL
- Componenti:Ferro
- Simbolo:Fe
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



