Avantor® Hichrom HI-5 Basic, GC Columns
GC column, Capillare, 1,00 µm, 0,53 mm, 50 m
Catalog # HI03-53-100-50
Supplier: Avantor
Jump to:
Product Details & Documents
Max temperature limits may change depending on the film thickness.
Specifications
- Formato della colonna:Capillare
- Spessore dello strato:1,00 µm
- Ø int.:0,53 mm
- Descrizione:GC column
- Lunghezza:50 m
- Cat. No.:HI03-53-100-50
- Crossbond:Yes
- Phase composition:5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- Polarity:Low-polarity stationary phase
- Phase name:HI-5 Basic
Accessories
Product Information
- Catalog #DescrizioneAvailabilityPrice
- HI0-100-PF-CLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLog in for availability1 * 100 ST - SFr. 806.00
Product Information
- Catalog #DescrizioneAvailabilityPrice
- HI0-10-PF-CLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLog in for availability1 * 10 ST - SFr. 111.00
Product Information
- Catalog #DescrizioneAvailabilityPrice
- HI0-10-PFY-C3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLog in for availability1 * 10 ST - SFr. 961.00
Product Information
- Catalog #DescrizioneAvailabilityPrice
- HI0-1-PFY-C3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLog in for availability1 * 1 ST - SFr. 99.40
Product Information
- Catalog #DescrizioneAvailabilityPrice
- HI0-25-PF-CLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLog in for availability1 * 25 ST - SFr. 215.00
Product Information
- Catalog #DescrizioneAvailabilityPrice
- HI0-2-PFY-C3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLog in for availability1 * 2 ST - SFr. 199.00
Product Information
- Catalog #DescrizioneAvailabilityPrice
- HI0-50-PF-CLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLog in for availability1 * 50 ST - SFr. 414.00
Product Information
- Catalog #DescrizioneAvailabilityPrice
- HI0-5-PF-CLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLog in for availability1 * 5 ST - SFr. 74.10
Product Information
- Catalog #DescrizioneAvailabilityPrice
- HI0-5-PFY-C3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLog in for availability1 * 5 ST - SFr. 491.00