Accessories for PLATEMASTER®
Height stop
N° de catalogue 631-1373
Fournisseur: GILSON
Aller à
Product Details & Documents
The Pipetting Head Height Adjuster is an accessory for the PLATEMASTER® that allows for easy altering of pipetting height, ensuring appropriate tip depth within shallow- or deep-well microplates.
The Lubrication Box of O-rings is an accessory for the PLATEMASTER®. We recommend lightly lubricating the external O-ring between regular maintenance intervals in situations of intensive use and to avoid ill-fitting tips.
Documents
Spécifications
- Descrizione:Height stop
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.