Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Pre-packed columns, 1 and 5 ml, SkillPak™

Pre-packed columns, 1 and 5 ml, SkillPak™

TOYOPEARL GigaCap CM-650M, Scouting column, 100 nm, 75 µm, 8,00 mm, 100 mm, 5 ml, 1,3 ml/min (max. 5 ml/min)
Codice catalogo TOYO0045247
Nome del fornitore:   TOSOH Bioscience
Pre-packed columns, 1 and 5 ml, SkillPak™
Codice catalogo TOYO0045247
Nome del fornitore:  TOSOH Bioscience
Pre-packed columns, 1 and 5 ml, SkillPak™
Pre-packed columns, 1 and 5 ml, SkillPak™

TOYOPEARL GigaCap CM-650M, Scouting column, 100 nm, 75 µm, 8,00 mm, 100 mm, 5 ml, 1,3 ml/min (max. 5 ml/min)

Price
294.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents

SkillPak 1 and 5 ml columns are designed for fast method development, resin screening or sample concentration. Pre-packed with TOYOPEARL®, TSKgel® or Ca++Pure-HA™ process chromatography media, these columns offer superior separation of biomelocules, such as monoclonal antibodies, proteins and oligonucleotides. SkillPak 50 and 200 ml pre-packed columns are the best solution for seamless scale up of methods developed on SkillPak 1 and 5.

  • Standard fittings (10-32 for 1/16" capillary)
  • Max. pressure: 3 bar (1 ml columns and 5 ml TOYOPEARL), ≤4 bar (5 ml TSKgel and Ca++Pure-HA)

SkillPak pre-packed columns are designed for your purification and separation tasks from platform design to pilot scale. These columns are ready to use upon receipt and show excellent physical strength and ideal flow characteristics for industrial downstream process.

These columns guarantee optimal performance and can be operated with commonly used low or media pressure liquid chromatography systems. They are reproducibly packed and take into account the varying compressibility of each resin. This provides an accurate representation of conditions found in full scale columns.

1 ml columns: Shipping buffer of 20% ethanol for TOYOPEARL and TSKgel (with the exception of 0,5 mol/L sodium citrate with 20% ethanol for TSKgel SP-5PW (20) and SuperQ-5PW (20)), 20 mmol/L phosphate with 20% ethanol for Ca++Pure-HA.

5 ml columns: Shipping buffer of 20% ethanol for TOYOPEARL and TSKgel, 20 mmol/L phosphate with 20% ethanol for Ca++Pure-HA. Asymmetry factor (As): 0,8 to 1,4 for TOYOPEARL and TSKgel, 0,8 to 2,6 for Ca++Pure-HA.

Specifiche
  • Formato della colonna:
    Scouting column
  • Velocità del flusso:
    1,3 ml/min (max. 5 ml/min)
  • Ø int.:
    8,00 mm
  • Dimensioni particelle:
    75 µm
  • Dimensioni pori:
    100 nm
  • Capacità:
    5 ml
  • Descrizione:
    TOYOPEARL GigaCap CM-650M
  • Lunghezza:
    100 mm