Orders LinkContactUsLinkComponent
Neomycin, Reference Standards
Neomycin, Reference Standards
Neomycin Sulfate System Suitability Mixture, USP reference standard, Vial, 100 mg
Codice catalogo 1458031.
Nome del fornitore:   USP
Neomycin, Reference Standards
Codice catalogo 1458031.
Nome del fornitore:  USP
Numero CAS:  
Neomycin, Reference Standards
Neomycin Sulfate System Suitability Mixture, USP reference standard, Vial, 100 mg
Numero CAS:  
Price
SFr. 516.00
Log in per disponibilità
1 * 100 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Documents
Specifiche
Specifiche
  • Tipo di conf.:
    Vial
  • Conf:
    100 mg
  • Standard:
    USP reference standard
  • Descrizione:
    Neomycin Sulfate System Suitability Mixture