Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Conjugated Linoleic Acids, Reference Standards

Conjugated Linoleic Acids, Reference Standards

Conjugated Linoleic Acids - Free Fatty Acid, USP reference standard, 3 x 100 mg, Ampule, 300 mg
Codice catalogo 1367676.
Nome del fornitore:   USP
Conjugated Linoleic Acids, Reference Standards
Codice catalogo 1367676.
Nome del fornitore:  USP
Numero CAS:  
Conjugated Linoleic Acids, Reference Standards

Conjugated Linoleic Acids - Free Fatty Acid, USP reference standard, 3 x 100 mg, Ampule, 300 mg

Numero CAS:  
Price
SFr. 532.00
Log in per disponibilità
1 * 300 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifiche

Specifiche

  • Tipo di conf.:
    Ampule
  • Conf:
    300 mg
  • Standard:
    USP reference standard
  • Descrizione:
    Conjugated Linoleic Acids - Free Fatty Acid
  • Pk Info:
    3 x 100 mg

Specifiche

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..