Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Conjugated Linoleic Acids, Reference Standards

Conjugated Linoleic Acids, Reference Standards

Conjugated Linoleic Acids - Triglycerides, USP reference standard, Ampule, 100 mg
Codice catalogo 1357090.
Nome del fornitore:   USP
Conjugated Linoleic Acids, Reference Standards
Codice catalogo 1357090.
Nome del fornitore:  USP
Numero CAS:  
Conjugated Linoleic Acids, Reference Standards

Conjugated Linoleic Acids - Triglycerides, USP reference standard, Ampule, 100 mg

Numero CAS:  
Price
481.00 SFr.
Log in per disponibilità
1 * 100 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifiche

Specifiche

  • Tipo di conf.:
    Ampule
  • Conf:
    100 mg
  • Standard:
    USP reference standard
  • Descrizione:
    Conjugated Linoleic Acids - Triglycerides

Specifiche

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..