Orders LinkContactUsLinkComponent
Castor Oil, Reference Standards
Castor Oil, Reference Standards
Castor Oil, USP reference standard, 3 x 100 mg, Ampule, 300 mg
Codice catalogo 1096826.
Nome del fornitore:   USP
Castor Oil, Reference Standards
Codice catalogo 1096826.
Nome del fornitore:  USP
Numero CAS:  
Castor Oil, Reference Standards
Castor Oil, USP reference standard, 3 x 100 mg, Ampule, 300 mg
Numero CAS:  
Price
SFr. 419.00
Log in per disponibilità
1 * 300 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Castor Oil: Mixture containing about 92% of Triricinolein
Zertifizierung: US Pharmacopea Reference Standard
Specifiche
Specifiche
  • Tipo di conf.:
    Ampule
  • Conf:
    300 mg
  • Standard:
    USP reference standard
  • Descrizione:
    Castor Oil
  • Pk Info:
    3 x 100 mg
  • Cat. No.:
    1096826.
  • Descrizione:
    Castor Oil
  • CAS No.:
    8001-79-4
Specifiche