ContactUsLinkComponent
FITC-Labeled Human EGF R, His Tag (EGR-HF2H5) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession P00533-1). It is the FITC labeled form of Human EGF R, His Tag (EGR-H5222). 1 * 200 µG
Codice catalogo ACRBEGR-HF2H5200UG
Nome del fornitore:  ACROBIOSYSTEMS
FITC-Labeled Human EGF R, His Tag (EGR-HF2H5) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession P00533-1). It is the FITC labeled form of Human EGF R, His Tag (EGR-H5222). 1 * 200 µG
Codice catalogo ACRBEGR-HF2H5200UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together