ContactUsLinkComponent
Recombinat Protein,Human E-Selectin CD62E Protein, Fc Tag,Human E-Selectin, Fc Tag (CDE-H5250) is expressed from human 293 cells (HE K293). It contains AA Trp 22 - Phe 556 (Accession NP000441.2).This protein carries a human IgG1 Fc tag at the C-termi 1 * 100 µG
Codice catalogo ACRBCDE-H5250100UG
Nome del fornitore:  ACRO Biosystems
Recombinat Protein,Human E-Selectin CD62E Protein, Fc Tag,Human E-Selectin, Fc Tag (CDE-H5250) is expressed from human 293 cells (HE K293). It contains AA Trp 22 - Phe 556 (Accession NP000441.2).This protein carries a human IgG1 Fc tag at the C-termi 1 * 100 µG
Codice catalogo ACRBCDE-H5250100UG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together