Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Human recombinant CD48 (from HEK293)

Human recombinant CD48 (from HEK293)

1 mg
Codice catalogo ACRBBC1-H5255-1MG
Marketsource
Nome del fornitore:   ACRO Biosystems
Human recombinant CD48 (from HEK293)
Codice catalogo ACRBBC1-H5255-1MG
Nome del fornitore:  ACRO Biosystems
Marketsource
Numero CAS:  
Human recombinant CD48 (from HEK293)

1 mg

Numero CAS:  
Price
SFr. 2’165.00
Log in per disponibilità
1 * 1 mg
Product Details & Documents
Specifiche

Specifiche

  • Conf:
    1 mg
  • Protease-free:
    Y
  • Senza animali:
    Y
  • Protein/peptide name:
    CD48
  • Purezza:
    >95%
  • Attività biologica:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human 2B4, His Tag (Cat. No. 2B4-H5224) at 2 μg/ml (100 μl/well) can bind Human CD48, Fc Tag (Cat. No. BC1-H5255) with a linear range of 0,078 - 2,5 μg/ml (QC tested). Measured by its binding ability in a functional ELISA. Immobilised Biotinylated Human 2B4, Avitag,His Tag (recommended for biopanning) (Cat. No. 2B4-H82E9) at 1 μg/ml (100 μl/well) on streptavidin precoated (0,2 μg/well) plate, can bind Human CD48, Fc Tag (Cat. No. BC1-H5255) with a linear range of 0,3 - 5 ng/ml (Routinely tested).
  • Protein synonyms:
    CD48|BCM-1|MEM-102|BLAST|SLAMF-2|BLAST1|BCM1|SLAMF2|BLAST-1|TCT.1
  • Specie:
    Human
  • Coniugazione:
    Non coniugato
  • Senza corriere:
    Y
  • Sorgente luminosa:
    HEK293 cells
  • Peso molecolare:
    49,0 kDa
  • Esente da nucleasi:
    Y
  • Cat. No.:
    ACRBBC1-H5255-1MG
  • Condizioni di conservazione:
    –20 °C
  • Contenuto di endotossine:
    Less than 1,0 EU per μg by the LAL method.
  • Protein/peptide type:
    Recombinant
  • Livello di endotossine:
    Low
  • ID gene:
    P09326-1
  • Formulazione:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Temperatura di spedizione:
    Powder, RT

Specifiche