ContactUsLinkComponent
MABSol® Biotinylated Mouse PD-L1, Fc,Avitag (PD1-M82F5) is expressed from human HEK293 cells. It contains AA Phe 19 - Thr 238 (Acces sion NP068693). 1 * 25 µG
Codice catalogo ACRBPD1-M82F5-25UG
Nome del fornitore:  ACRO Biosystems
MABSol® Biotinylated Mouse PD-L1, Fc,Avitag (PD1-M82F5) is expressed from human HEK293 cells. It contains AA Phe 19 - Thr 238 (Acces sion NP068693). 1 * 25 µG
Codice catalogo ACRBPD1-M82F5-25UG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together