Cynomolgus Rhesus macaque BCMA, Fc Tag (BCA-C5253) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Access ion G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are homologus. 1 * 1 mg
Catalog # ACRBBCA-C5253-1MG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together