ContactUsLinkComponent
Recombinat Protein,Human Activin RIIB ACVR2B Protein, His Tag,Human Activin RIIB, His Tag (ACB-H5226) is expressed from human 293 ce lls (HEK293). It contains AA Ser 19 - Thr 137 (Accession NP001097).This protein carries a polyhistidine tag at the C- 1 * 200 µG
Codice catalogo ACRBACB-H5226200UG
Nome del fornitore:  ACROBIOSYSTEMS
Recombinat Protein,Human Activin RIIB ACVR2B Protein, His Tag,Human Activin RIIB, His Tag (ACB-H5226) is expressed from human 293 ce lls (HEK293). It contains AA Ser 19 - Thr 137 (Accession NP001097).This protein carries a polyhistidine tag at the C- 1 * 200 µG
Codice catalogo ACRBACB-H5226200UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together