Recombinat Protein,Cynomolgus LILRB4 CD85k ILT3 Protein, Fc Tag,Cynomolgus LILRB4, Fc Tag (CDK-C5258) is expressed from human 293 ce lls (HEK293). It contains AA Gln 22 - Glu 259 (Accession XP015297198.1).This protein carries a human IgG1 Fc tag at t 1 * 100 µG
Codice catalogo ACRBCDK-C5258100UG
Nome del fornitore: ACRO Biosystems
Frequently Bought Together