ContactUsLinkComponent
Recombinat Protein,Human Angiopoietin-like 4 ANGPTL4 (166-406) Protein, His Tag,Human Angiopoietin-like 4 (166-406), His Tag (AN4-H5 243) is expressed from human 293 cells (HEK293). It contains AA Pro 166 - Ser 406 (Accession NP647475).This protein c 1 * 1 mg
Catalog # ACRBAN4-H5243-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human Angiopoietin-like 4 ANGPTL4 (166-406) Protein, His Tag,Human Angiopoietin-like 4 (166-406), His Tag (AN4-H5 243) is expressed from human 293 cells (HEK293). It contains AA Pro 166 - Ser 406 (Accession NP647475).This protein c 1 * 1 mg
Catalog # ACRBAN4-H5243-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together