Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Mouse recombinant B7-H5 (from HEK293)

100 µG
  ACRBB75-M52H7100UG
Marketsource
Mouse recombinant B7-H5 (from HEK293)
  ACRBB75-M52H7100UG
Marketsource
 :  
Mouse recombinant B7-H5 (from HEK293)

100 µG

 :  
 
SFr. 360.00
   
1 * 100 µG
 
 

 

  • Conf:
    100 µG
  • Protease-free:
    Y
  • Senza animali:
    Y
  • Protein/peptide name:
    B7-H5
  • Purezza:
    >95%
  • Attività biologica:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Protein synonyms:
    B7-H5|SISP1|Gi24|VISTA
  • Specie:
    Mouse
  • Coniugazione:
    Non coniugato
  • Senza corriere:
    Y
  • Sorgente luminosa:
    HEK293 cells
  • Peso molecolare:
    19,7kDa
  • Esente da nucleasi:
    Y
  • Cat. No.:
    ACRBB75-M52H7100UG
  • Condizioni di conservazione:
    –20 °C
  • Contenuto di endotossine:
    Less than 1,0 EU per μg by the LAL method
  • Protein/peptide type:
    Recombinant
  • Livello di endotossine:
    Low
  • ID gene:
    Q9D659-1
  • Formulazione:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Temperatura di spedizione:
    Powder, RT