Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Mouse recombinant BAFFR (from HEK293)

Mouse recombinant BAFFR (from HEK293)

1 mg
Catalog # ACRBBAR-M5259-1MG
Marketsource
Supplier:   ACRO Biosystems
Mouse recombinant BAFFR (from HEK293)
Catalog # ACRBBAR-M5259-1MG
Supplier:  ACRO Biosystems
Marketsource
CAS Number:  
Mouse recombinant BAFFR (from HEK293)

1 mg

CAS Number:  
Price
SFr. 2’495.00
Log in for availability
1 * 1 mg
Product Details & Documents
Specifications

Specifications

  • Conf:
    1 mg
  • Protease-free:
    Y
  • Senza animali:
    Y
  • Protein/peptide name:
    BAFFR
  • Purezza:
    >95%
  • Attività biologica:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
  • Protein synonyms:
    CVID4|BAFFR|BAFF-R|BROMIX|CD268|TNFRSF13C|prolixin
  • Specie:
    Mouse
  • Coniugazione:
    Non coniugato
  • Senza corriere:
    Y
  • Sorgente luminosa:
    HEK293 cells
  • Peso molecolare:
    33,3 kDa
  • Esente da nucleasi:
    Y
  • Cat. No.:
    ACRBBAR-M5259-1MG
  • Condizioni di conservazione:
    –20 °C
  • Contenuto di endotossine:
    Less than 1,0 EU per μg by the LAL method
  • Protein/peptide type:
    Recombinant
  • Livello di endotossine:
    Low
  • ID gene:
    Q9D8D0
  • Formulazione:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
  • Temperatura di spedizione:
    Powder, RT

Specifications