Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 SFr.
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Human CD117, His Tag (CD7-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Thr 516 (Accession P10721-2). 1 * 100 µG
Catalog # ACRBCD7-H52H4100UG
Supplier:
ACROBIOSYSTEMS
Expand image
Human CD117, His Tag (CD7-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Thr 516 (Accession P10721-2). 1 * 100 µG
Catalog # ACRBCD7-H52H4100UG
Supplier:
ACROBIOSYSTEMS
Frequently Bought Together
Pasteur pipettes, disposable
Supplier: Brand
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Pipette sierologiche, Standard Line
Supplier: VWR Collection
Provette per centrifuga, Falcon®
Supplier: Corning
VWR®, contenitori per campioni conici, con coperchi a scatto
Supplier: VWR Collection
VWR®, Pipette Pasteur, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Supplier: VWR Collection
Centrifuge tubes, Falcon®
Supplier: Corning
VWR®, CryoBox, cartone, 133×133 mm
Supplier: VWR Collection
VWR®, pipette di trasferimento, alte prestazioni
Supplier: VWR Collection
VWR®, Piastra Petri, PS
Supplier: VWR Collection
VWR®, Contenitori per campioni dritti con tappo a vite
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF