ContactUsLinkComponent
Biotinylated Human 4-1BB Ligand, Fc,Avitag (41L-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 50 - Glu 254 ( Accession P41273-1). 1 * 25 µG
Catalog # ACRB41L-H82F9-25UG
Supplier:  ACRO Biosystems
Biotinylated Human 4-1BB Ligand, Fc,Avitag (41L-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 50 - Glu 254 ( Accession P41273-1). 1 * 25 µG
Catalog # ACRB41L-H82F9-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together