Human LAG-3, Fc Tag (HPLC-verified) (LA3-H5255) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accessi on P18627-1). 1 * 100 µG
Codice catalogo ACRBLA3-H5255100UG
Nome del fornitore: ACRO Biosystems
Frequently Bought Together