Biotinylated Mouse OX40, His,Avitag (OX0-M82E5) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 211 (Accessi on P47741-1). 1 * 25 µG
Codice catalogo ACRBOX0-M82E5-25UG
Nome del fornitore: ACRO Biosystems
Frequently Bought Together