Biotinylated HIV-1 HIV-1Clade BC (CN54) GP120, His,Avitag derived from the env. gene of HIV-1 strain CN54 gp160 (Accession G4XFJ5-1 (E46G, T396A, A497T), Thr 36 - Arg 507) and glycosylated with N-linked sugars and expressed in HEK293 cells at ACRObio 1 * 25 µG
Codice catalogo ACRBGP0-V182E625UG
Nome del fornitore: ACROBIOSYSTEMS
Frequently Bought Together