ContactUsLinkComponent
Human Siglec-9, His Tag (SI9-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Gly 348 (Accession Q9Y336-1) . 1 * 100 µG
Codice catalogo ACRBSI9-H52H4100UG
Nome del fornitore:  ACROBIOSYSTEMS
Human Siglec-9, His Tag (SI9-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Gly 348 (Accession Q9Y336-1) . 1 * 100 µG
Codice catalogo ACRBSI9-H52H4100UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together