ContactUsLinkComponent
Human Siglec-9, Fc Tag (SI9-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Gly 348 (Accession Q9Y336-1). 1 * 1 mg
Catalog # ACRBSI9-H5256-1MG
Supplier:  ACROBIOSYSTEMS
Human Siglec-9, Fc Tag (SI9-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 18 - Gly 348 (Accession Q9Y336-1). 1 * 1 mg
Catalog # ACRBSI9-H5256-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together