Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Human Her2, Tag Free (HE2-H5212) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession P04626-1). 1 * 50 µG
ACRBHE2-H5212-50UG
:
ACRO Biosystems
Human Her2, Tag Free (HE2-H5212) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession P04626-1). 1 * 50 µG
ACRBHE2-H5212-50UG
:
ACRO Biosystems
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acido cloridrico 1 mol/l (1 N), Titripur® Reag. USP, Reag. Ph. Eur. soluzione volumetrica, Supelco®
: Merck
Piastre per microtitolazione a 96 e 384 pozzetti, Nunc™
: Thermo Fisher Scientific
Sodio cloruro, EMSURE® ACS, ISO, Reag. Ph. Eur. per analisi, Supelco®
: Merck
VWR®, pipette di trasferimento, alte prestazioni
: VWR Collection
VWR® NITRILE LIGHT, Guanti in nitrile
: VWR Collection
VWR®, Pipette sierologiche, Standard Line
: VWR Collection
VWR®, Guanti in Nitrile
: VWR Collection
VWR®, Salviette per il viso
: VWR Collection
Unità di filtrazione sottovuoto, filtri Rapid-Flow™ MF 75, Nalgene®
: Thermo Fisher Scientific
Strisce reattive pH, MQuant®, Supelco®
: Merck
VWR®, Pipette Pasteur, Standard Line
: VWR Collection
VWR®, pellicole in poliestere per ELISA e incubazione generale
: VWR Collection
Guanti in nitrile, Microflex® 93-853
: Ansell
ADENINE (HEMISULFATE) 1 * 10 g
: Cayman Chemical
VWR®, Vaschette per reagenti
: VWR Collection