Biotinylated Human IL-2 R alpha, His,Avitag (ILA-H82E6) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession P01589-1). 1 * 25 µG
Biotinylated Human IL-2 R alpha, His,Avitag (ILA-H82E6) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession P01589-1). 1 * 25 µG