ContactUsLinkComponent
Biotinylated Human HGF R, Avitag,His Tag (MET-H82E1) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Thr 932 (Ac cession AAI30421.1). 1 * 25 µG
Codice catalogo ACRBMET-H82E1-25UG
Nome del fornitore:  ACRO Biosystems
Biotinylated Human HGF R, Avitag,His Tag (MET-H82E1) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Thr 932 (Ac cession AAI30421.1). 1 * 25 µG
Codice catalogo ACRBMET-H82E1-25UG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together