Mouse Complement C5, His Tag (CO5-M52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 1680 (Accession P06 684-1). 1 * 100 µG
Codice catalogo ACRBCO5-M52H4100UG
Nome del fornitore: ACRO Biosystems
Frequently Bought Together