ContactUsLinkComponent
Human CD96, Mouse IgG2a Fc Tag, low endotoxin (TAE-H5252) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Met 50 3 (Accession P40200-2). 1 * 100 µG
Codice catalogo ACRBTAE-H5252100UG
Nome del fornitore:  ACRO Biosystems
Human CD96, Mouse IgG2a Fc Tag, low endotoxin (TAE-H5252) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Met 50 3 (Accession P40200-2). 1 * 100 µG
Codice catalogo ACRBTAE-H5252100UG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together