ContactUsLinkComponent
Human GITR Ligand, Fc Tag (GIL-H526a) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession AAH6931 9.1). 1 * 50 µG
Codice catalogo ACRBGIL-H526A-50UG
Nome del fornitore:  ACROBIOSYSTEMS
Human GITR Ligand, Fc Tag (GIL-H526a) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession AAH6931 9.1). 1 * 50 µG
Codice catalogo ACRBGIL-H526A-50UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together