ContactUsLinkComponent
Cynomolgus Siglec-15, His Tag (SG5-C52H6) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Accession A0A 2K5UY47-1). 1 * 1 mg
Codice catalogo ACRBSG5-C52H6-1MG
Nome del fornitore:  ACRO Biosystems
Cynomolgus Siglec-15, His Tag (SG5-C52H6) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Accession A0A 2K5UY47-1). 1 * 1 mg
Codice catalogo ACRBSG5-C52H6-1MG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together