FITC-Labeled Human Mesothelin (296-580), His Tag (Cat. No. MSN-HF223) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession AAH09272). It is the FITC labeled form of Human Mesothelin (296-580), His Tag (Cat. No. MSN-H5223 1 * 25 µG
Codice catalogo ACRBMSN-HF223-25UG
Nome del fornitore: ACROBIOSYSTEMS
Frequently Bought Together