Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Human recombinant integrin alpha 4 beta 1 (from HEK293)

Human recombinant integrin alpha 4 beta 1 (from HEK293)

25 µG
Codice catalogo ACRBIT1-H82W1-25UG
Marketsource
Nome del fornitore:   ACRO Biosystems
Human recombinant integrin alpha 4 beta 1 (from HEK293)
Codice catalogo ACRBIT1-H82W1-25UG
Nome del fornitore:  ACRO Biosystems
Marketsource
Numero CAS:  
Human recombinant integrin alpha 4 beta 1 (from HEK293)

25 µG

Numero CAS:  
Price
SFr. 360.00
Log in per disponibilità
1 * 25 µG
Product Details & Documents
Specifiche

Specifiche

  • Conf:
    25 µG
  • Protease-free:
    Y
  • Senza animali:
    Y
  • Protein/peptide name:
    Integrin alpha 4 beta 1
  • Purezza:
    >95%
  • Attività biologica:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised natalizumab at 1 μg/ml (100 μl/well) can bind biotinylated human ITGA4 and ITGB1 heterodimer protein, His,Avitag and tag free (Cat. No. ACRBIT1-H82W1) with a linear range of 2 - 31 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised Human VCAM-1, His Tag (Cat. No. ACRBVC1-H5224) at 5 μg/ml (100 μl/well) can bind biotinylated human ITGA4 and ITGB1 heterodimer protein, His,Avitag and tag free (Cat. No. ACRBIT1-H82W1) with a linear range of 0,01 - 0,156 μg/ml (routinely tested).
  • Protein synonyms:
    Integrin alpha 4 beta 1|ITGA4 and ITGB1
  • Specie:
    Human
  • Senza corriere:
    Y
  • Sorgente luminosa:
    HEK293 cells
  • Peso molecolare:
    113,2 kDa (ITGA4) and 83,7 kDa (ITGB1)
  • Esente da nucleasi:
    Y
  • Cat. No.:
    ACRBIT1-H82W1-25UG
  • Condizioni di conservazione:
    –20 °C
  • Contenuto di endotossine:
    Less than 1,0 EU per μg by the LAL method.
  • Protein/peptide type:
    Recombinant
  • Livello di endotossine:
    Low
  • ID gene:
    P13612-1 (ITGA4) and P05556-1 (ITGB1)
  • Formulazione:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Temperatura di spedizione:
    Powder,RT

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.