Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Human recombinant integrin alpha 4 beta 1 (from HEK293)

Human recombinant integrin alpha 4 beta 1 (from HEK293)

200 µG
Catalog # ACRBIT1-H82W1200UG
Marketsource
Supplier:   ACRO Biosystems
Human recombinant integrin alpha 4 beta 1 (from HEK293)
Catalog # ACRBIT1-H82W1200UG
Supplier:  ACRO Biosystems
Marketsource
CAS Number:  
Human recombinant integrin alpha 4 beta 1 (from HEK293)

200 µG

CAS Number:  
Price
SFr. 1’565.00
Log in for availability
1 * 200 µG
Product Details & Documents
Specifications

Specifications

  • Conf:
    200 µG
  • Protease-free:
    Y
  • Senza animali:
    Y
  • Protein/peptide name:
    Integrin alpha 4 beta 1
  • Purezza:
    >95%
  • Attività biologica:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised natalizumab at 1 μg/ml (100 μl/well) can bind biotinylated human ITGA4 and ITGB1 heterodimer protein, His,Avitag and tag free (Cat. No. ACRBIT1-H82W1) with a linear range of 2 - 31 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised Human VCAM-1, His Tag (Cat. No. ACRBVC1-H5224) at 5 μg/ml (100 μl/well) can bind biotinylated human ITGA4 and ITGB1 heterodimer protein, His,Avitag and tag free (Cat. No. ACRBIT1-H82W1) with a linear range of 0,01 - 0,156 μg/ml (routinely tested).
  • Protein synonyms:
    Integrin alpha 4 beta 1|ITGA4 and ITGB1
  • Specie:
    Human
  • Senza corriere:
    Y
  • Sorgente luminosa:
    HEK293 cells
  • Peso molecolare:
    113,2 kDa (ITGA4) and 83,7 kDa (ITGB1)
  • Esente da nucleasi:
    Y
  • Cat. No.:
    ACRBIT1-H82W1200UG
  • Condizioni di conservazione:
    –20 °C
  • Contenuto di endotossine:
    Less than 1,0 EU per μg by the LAL method.
  • Protein/peptide type:
    Recombinant
  • Livello di endotossine:
    Low
  • ID gene:
    P13612-1 (ITGA4) and P05556-1 (ITGB1)
  • Formulazione:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Temperatura di spedizione:
    Powder,RT

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.