Biotinylated Human GITR Ligand, Avitag,Fc Tag (GIL-H82F8) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 17 7 (Accession AAH69319.1). 1 * 25 µG
Codice catalogo ACRBGIL-H82F8-25UG
Nome del fornitore: ACROBIOSYSTEMS
Frequently Bought Together