Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Human FOLR2, His Tag (FO2-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 17 - His 228 (Accession P14207-1). 1 * 1 mg
ACRBFO2-H5223-1MG
:
ACRO Biosystems
Human FOLR2, His Tag (FO2-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 17 - His 228 (Accession P14207-1). 1 * 1 mg
ACRBFO2-H5223-1MG
:
ACRO Biosystems
Frequently Bought Together
VWR®, Piastre multipozzetto per colture cellulari
: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR®, Pipette sierologiche, Standard Line
: VWR Collection
VWR®, Sacchetti per omogenizzatore
: VWR Collection
Pipette sierologiche, Stripette™
: Corning
VWR®, CryoBox, cartone, 133×133 mm
: VWR Collection
VWR®, pipette di trasferimento, alte prestazioni
: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
: VWR Collection
VWR®, Contenitori per campioni dritti con tappo a vite
: VWR Collection
Centrifuge tubes, 15 ml, Corning®
: Corning
VWR®, Pipette Pasteur, Standard Line
: VWR Collection
Multiwell cell culture plates, Falcon®
: Corning
Pipette per sierologia
: Greiner Bio-One
Provetta per centrifuga con tappo a vite
: Greiner Bio-One
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
Provette per microcentrifuga Eppendorf® Protein LoBind®
: EPPENDORF
Human FOLR3, His Tag (FO3-H52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Ser 245 (Accession # P41439-1). 1 * 100 µG
: ACRO Biosystems