ContactUsLinkComponent
Human Siglec-3, His Tag (CD3-H5226) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession AAH28152. 1). 1 * 100 µG
Codice catalogo ACRBCD3-H5226100UG
Nome del fornitore:  ACROBIOSYSTEMS
Human Siglec-3, His Tag (CD3-H5226) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession AAH28152. 1). 1 * 100 µG
Codice catalogo ACRBCD3-H5226100UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together