ContactUsLinkComponent
Biotinylated Human FGL1, Avitag,Fc Tag (FG1-H82F4) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Acce ssion Q08830-1). 1 * 200 µG
Catalog # ACRBFG1-H82F4200UG
Supplier:  ACRO Biosystems
Biotinylated Human FGL1, Avitag,Fc Tag (FG1-H82F4) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Acce ssion Q08830-1). 1 * 200 µG
Catalog # ACRBFG1-H82F4200UG
Supplier:  ACRO Biosystems
Frequently Bought Together