ContactUsLinkComponent
Recombinat Protein,Human TIMP1 Protein, His Tag,Human TIMP1, His Tag (TI1-H5226) is expressed from human 293 cells (HEK293). It cont ains AA Cys 24 - Ala 207 (Accession AAH00866).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 1 mg
  ACRBTI1-H5226-1MG
Recombinat Protein,Human TIMP1 Protein, His Tag,Human TIMP1, His Tag (TI1-H5226) is expressed from human 293 cells (HEK293). It cont ains AA Cys 24 - Ala 207 (Accession AAH00866).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 1 mg
  ACRBTI1-H5226-1MG
Frequently Bought Together