FITC-Labeled Human CD4, His Tag (CD4-HF2H7) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession A AH25782). It is the FITC labeled form of Human CD4, His Tag (LE3-H5228). 1 * 25 µG
Codice catalogo ACRBCD4-HF2H7-25UG
Nome del fornitore: ACROBIOSYSTEMS
Frequently Bought Together