ContactUsLinkComponent
Biotinylated Human Mucin-1 (890-1158), His,Avitag (MU1-H82E5) is expressed from human 293 cells (HEK293). It contains AA Ser 890 - G ly 1158 (Accession P15941-1). 1 * 200 µG
Catalog # ACRBMU1-H82E5200UG
Supplier:  ACRO Biosystems
Biotinylated Human Mucin-1 (890-1158), His,Avitag (MU1-H82E5) is expressed from human 293 cells (HEK293). It contains AA Ser 890 - G ly 1158 (Accession P15941-1). 1 * 200 µG
Catalog # ACRBMU1-H82E5200UG
Supplier:  ACRO Biosystems
Frequently Bought Together