Recombinat Protein,Human R-Spondin 1 RSPO1 (21-146) Protein, His Tag,Human R-Spondin 1 (21-146), His Tag (RS6-H4220) is expressed fr om human 293 cells (HEK293). It contains AA Ser 21 - Ala 146 (Accession AAI14967).This protein carries a polyhistidin 1 * 1 mg
Catalog # ACRBRS6-H4220-1MG
Supplier: ACRO Biosystems
Frequently Bought Together